1-(3-(Cyclopropylmethoxy)-4-methylphenyl)propan-1-one ≥95%

Supplier: AOBChem USA

Warning

515049-5G 515049-25G 515049-1G 515049-500MG
77630-164EA 1484.23 USD
77630-164 77656-374 77643-542 77651-482
1-(3-(Cyclopropylmethoxy)-4-methylphenyl)propan-1-one ≥95%
1-(3-(Cyclopropylmethoxy)-4-methylphenyl)propan-1-one

Precaución: For Research Use Only

Formula: 218.292 g/mol
MW: C₁₄H₁₈O₂
Storage Temperature: Ambient
CAS Number: 2162906-57-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR