2-(3-(Cyclopropylmethoxy)-4-methylphenyl)propan-2-ol ≥95%

Supplier: AOBChem USA

Warning

515048-5G 515048-25G 515048-1G 515048-500MG
77630-162EA 1484.23 USD
77630-162 77656-372 77643-540 77651-480
2-(3-(Cyclopropylmethoxy)-4-methylphenyl)propan-2-ol ≥95%
2-(3-(Cyclopropylmethoxy)-4-methylphenyl)propan-2-ol

Precaución: For Research Use Only

Formula: 220.307 g/mol
MW: C₁₄H₂₀O₂
Storage Temperature: Ambient
CAS Number: 3037692-93-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR