Ethyl 2-(2,3-difluoro-4-iodophenyl)-2-oxoacetate ≥97%

Supplier: AOBChem USA

Warning

515041-5G 515041-25G 515041-1G 515041-500MG
77630-160EA 1484.23 USD
77630-160 77636-062 77639-494 77647-480
Ethyl 2-(2,3-difluoro-4-iodophenyl)-2-oxoacetate ≥97%
Ethyl 2-(2,3-difluoro-4-iodophenyl)-2-oxoacetate

Precaución: For Research Use Only

Formula: 340.062 g/mol
MW: C₁₀H₇F₂IO₃
Storage Temperature: Ambient
CAS Number: 3037696-19-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR