1-(2,3-Difluoro-4-(methylthio)phenyl)cyclobutanol ≥95%

Supplier: AOBChem USA

Warning

515039-5G 515039-1G 515039-25G 515039-500MG
77630-158EA 1484.23 USD
77630-158 77639-492 77659-902 77651-478
1-(2,3-Difluoro-4-(methylthio)phenyl)cyclobutanol ≥95%
1-(2,3-Difluoro-4-(methylthio)phenyl)cyclobutanol

Precaución: For Research Use Only

Formula: C₁₁H₁₂F₂OS
Storage Temperature: Ambient
CAS Number: 3037703-95-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR