2-(Benzyloxy)-5-(trifluoromethyl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

51459-5G 51459-1G 51459-25G 51459-500MG
77635-134EA 586 USD
77635-134 77639-488 77656-366 77651-472
2-(Benzyloxy)-5-(trifluoromethyl)benzaldehyde ≥97%
2-(Benzyloxy)-5-(trifluoromethyl)benzaldehyde

Precaución: For Research Use Only

Formula: 280.242 g/mol
MW: C₁₅H₁₁F₃O₂
Storage Temperature: Ambient
MDL Number: MFCD29039640
CAS Number: 632626-08-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR