2-Bromo-6-chloro-4-isopropylphenol ≥97%

Supplier: AOBChem USA

Warning

51422-5G 51422-1G 51422-500MG
77659-898EA 2023 USD
77659-898 77643-534 77651-470
2-Bromo-6-chloro-4-isopropylphenol ≥97%
2-Bromo-6-chloro-4-isopropylphenol

Precaución: For Research Use Only

Formula: 249.532 g/mol
MW: C₉H₁₀BrClO
Storage Temperature: Ambient
CAS Number: 1784972-66-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR