2,4-Difluoro-3-methoxyphenol ≥97%

Supplier: AOBChem USA

Warning

51413-5G 51413-25G
77635-132EA 178 USD
77635-132 77659-896
2,4-Difluoro-3-methoxyphenol ≥97%
2,4-Difluoro-3-methoxyphenol

Precaución: For Research Use Only

Formula: C₇H₆F₂O₂
Storage Temperature: Ambient
MDL Number: MFCD04115938
CAS Number: 886499-27-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR