4-(4-Bromophenoxy)-1-methylpiperidine ≥97%

Supplier: AOBChem USA

Warning

513616-5G 513616-1G 513616-25G 513616-500MG
77633-488EA 1055 USD
77633-488 77639-486 77656-362 77647-476
4-(4-Bromophenoxy)-1-methylpiperidine ≥97%
4-(4-Bromophenoxy)-1-methylpiperidine

Precaución: For Research Use Only

Formula: 270.17 g/mol
MW: C₁₂H₁₆BrNO
Storage Temperature: Ambient
CAS Number: 494777-78-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR