4-(2-Bromophenoxy)-1-methylpiperidine ≥97%
Supplier: AOBChem USA
|
Warning
|
513609-5G
513609-1G
513609-25G
513609-500MG
77633-486EA
1055
USD
77633-486
77639-484
77659-894
77651-468
4-(2-Bromophenoxy)-1-methylpiperidine ≥97%
4-(2-Bromophenoxy)-1-methylpiperidine
Precaución: For Research Use Only
Formula:
270.17 g/mol MW: C₁₂H₁₆BrNO Storage Temperature: Ambient |
CAS Number:
494774-24-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...