5-Chloro-2-phenoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

513263-5G 513263-25G 513263-1G 513263-500MG
77630-152EA 1484.23 USD
77630-152 77636-060 77643-530 77651-464
5-Chloro-2-phenoxybenzaldehyde ≥97%
5-Chloro-2-phenoxybenzaldehyde

Precaución: For Research Use Only

Formula: 232.662 g/mol
MW: C₁₃H₉ClO₂
Storage Temperature: Ambient
CAS Number: 1290052-60-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR