Naphthalen-2-yl sulfofluoridate ≥97%

Supplier: AOBChem USA

Warning

51324-5G 51324-25G 51324-1G 51324-500MG
77633-484EA 988 USD
77633-484 77659-888 77643-528 77651-462
Naphthalen-2-yl sulfofluoridate ≥97%
Naphthalen-2-yl sulphofluoridate

Precaución: For Research Use Only

Formula: C₁₀H₇FO₃S
Storage Temperature: Ambient
MDL Number: MFCD29472040
CAS Number: 141694-39-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR