1-(3-Methyl-5-(methylthio)phenyl)cyclopentanol ≥95%

Supplier: AOBChem USA

Warning

513149-5G 513149-25G 513149-1G 513149-500MG
77630-146EA 1484.23 USD
77630-146 77656-358 77643-526 77651-460
1-(3-Methyl-5-(methylthio)phenyl)cyclopentanol ≥95%
1-(3-Methyl-5-(methylthio)phenyl)cyclopentanol

Precaución: For Research Use Only

Formula: 222.3464 g/mol
MW: C₁₃H₁₈OS
Storage Temperature: Ambient
CAS Number: 3037695-57-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR