(3-Chloro-2-fluoro-5-methylphenyl)(cyclopropyl)methanone ≥95%

Supplier: AOBChem USA

Warning

513061-5G 513061-25G 513061-1G 513061-500MG
77633-482EA 1441 USD
77633-482 77659-886 77643-524 77651-458
(3-Chloro-2-fluoro-5-methylphenyl)(cyclopropyl)methanone ≥95%
(3-Chloro-2-fluoro-5-methylphenyl)(cyclopropyl)methanone

Precaución: For Research Use Only

Formula: 212.648 g/mol
MW: C₁₁H₁₀ClFO
Storage Temperature: Ambient
CAS Number: 3037676-47-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR