3-(3-Chloro-2-fluoro-5-methylphenyl)oxetan-3-ol ≥95%

Supplier: AOBChem USA

Warning

513060-5G 513060-1G 513060-500MG
77656-356EA 1988 USD
77656-356 77643-522 77651-456
3-(3-Chloro-2-fluoro-5-methylphenyl)oxetan-3-ol ≥95%
3-(3-Chloro-2-fluoro-5-methylphenyl)oxetan-3-ol

Precaución: For Research Use Only

Formula: C₁₀H₁₀ClFO₂
Storage Temperature: Ambient
CAS Number: 3037676-36-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR