1-(3-Chloro-2-fluoro-5-methylphenyl)cyclobutanol ≥95%

Supplier: AOBChem USA

Warning

513059-1G 513059-5G 513059-500MG
77639-480EA 556 USD
77639-480 77656-354 77651-454
1-(3-Chloro-2-fluoro-5-methylphenyl)cyclobutanol ≥95%
1-(3-Chloro-2-fluoro-5-methylphenyl)cyclobutanol

Precaución: For Research Use Only

Formula: C₁₁H₁₂ClFO
Storage Temperature: Ambient
CAS Number: 3037703-16-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR