1-Chloro-3-ethyl-2-fluoro-5-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

513058-5G 513058-1G 513058-25G 513058-500MG
77630-144EA 1484.23 USD
77630-144 77639-478 77659-884 77651-452
1-Chloro-3-ethyl-2-fluoro-5-methylbenzene ≥95%
1-Chloro-3-ethyl-2-fluoro-5-methylbenzene

Precaución: For Research Use Only

Formula: 172.627 g/mol
MW: C₉H₁₀ClF
Storage Temperature: Ambient
CAS Number: 1807166-19-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR