2-(3-Chloro-2,4,6-trifluorophenyl)propan-2-ol ≥95%

Supplier: AOBChem USA

Warning

513055-5G 513055-1G 513055-500MG
77659-882EA 2053 USD
77659-882 77643-520 77647-466
2-(3-Chloro-2,4,6-trifluorophenyl)propan-2-ol ≥95%
2-(3-Chloro-2,4,6-trifluorophenyl)propan-2-ol

Precaución: For Research Use Only

Formula: C₉H₈ClF₃O
Storage Temperature: Ambient
CAS Number: 1892104-17-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR