(1-Methylenebutyl)benzene ≥95%

Supplier: AOBChem USA

Warning

512887-5G 512887-1G 512887-25G 512887-500MG
77633-480EA 1441 USD
77633-480 77639-474 77659-878 77651-450
(1-Methylenebutyl)benzene ≥95%
(1-Methylenebutyl)benzene

Precaución: For Research Use Only

Formula: C₁₁H₁₄
Storage Temperature: Ambient
CAS Number: 5676-32-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR