2-(4-Bromo-2-propoxyphenyl)-1,3-dioxolane ≥95%

Supplier: AOBChem USA

Warning

51237-5G 51237-25G 51237-1G 51237-500MG
77633-478EA 644 USD
77633-478 77656-352 77643-518 77651-448
2-(4-Bromo-2-propoxyphenyl)-1,3-dioxolane ≥95%
2-(4-Bromo-2-propoxyphenyl)-1,3-dioxolane

Precaución: For Research Use Only

Formula: 287.15 g/mol
MW: C₁₂H₁₅BrO₃
Storage Temperature: Ambient
MDL Number: MFCD34530073
CAS Number: 2734772-69-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR