5-(Methylthio)picolinic acid ≥95%

Supplier: AOBChem USA

Warning

51087-1G 51087-5G 51087-500MG
77639-472EA 571 USD
77639-472 77659-876 77647-462
5-(Methylthio)picolinic acid ≥95%
5-(Methylthio)picolinic acid

Precaución: For Research Use Only

Formula: 169.201 g/mol
MW: C₇H₇NO₂S
Storage Temperature: Ambient
MDL Number: MFCD00661386
CAS Number: 74470-29-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR