2-Methoxy-5-(methylthio)pyridine ≥95%

Supplier: AOBChem USA

Warning

51085-5G 51085-1G 51085-500MG
77630-140EA 1484.23 USD
77630-140 77643-514 77647-460
2-Methoxy-5-(methylthio)pyridine ≥95%
2-Methoxy-5-(methylthio)pyridine

Precaución: For Research Use Only

Formula: C₇H₉NOS
MW: 155.217 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28666740
CAS Number: 98627-15-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR