1-(4,5-Dichloropyridin-3-yl)-2,2,2-trifluoroethanone ≥95%
Supplier: AOBChem USA
|
Warning
|
50932-5G
50932-25G
50932-1G
50932-500MG
77630-134EA
1484.23
USD
77630-134
77656-350
77643-508
77647-456
1-(4,5-Dichloropyridin-3-yl)-2,2,2-trifluoroethanone ≥95%
1-(4,5-Dichloropyridin-3-yl)-2,2,2-trifluoroethanone
Precaución: For Research Use Only
Formula:
243.998 g/mol MW: C₇H₂Cl₂F₃NO Storage Temperature: Ambient |
MDL Number:
MFCD28137463 CAS Number: 1823320-66-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...