N-Methoxy-N-methylacrylamide ≥95%

Supplier: AOBChem USA

Warning

508552-5G 508552-1G 508552-500MG 508552-25G
77633-476EA 816 USD
77633-476 77639-466 77652-978 77656-346
N-Methoxy-N-methylacrylamide ≥95%
N-Methoxy-N-methylacrylamide

Precaución: For Research Use Only

Formula: C₅H₉NO₂
Storage Temperature: Ambient
MDL Number: MFCD12095636
CAS Number: 193634-77-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR