N-(2-Bromo-4,5-dimethoxyphenyl)acetamide ≥97%

Supplier: AOBChem USA

Warning

508547-5G 508547-25G 508547-1G 508547-500MG
77630-128EA 1583.11 USD
77630-128 77636-050 77639-462 77651-442
N-(2-Bromo-4,5-dimethoxyphenyl)acetamide ≥97%
N-(2-Bromo-4,5-dimethoxyphenyl)acetamide

Precaución: For Research Use Only

Formula: 274.111 g/mol
MW: C₁₀H₁₂BrNO₃
Storage Temperature: Ambient
CAS Number: 5349-12-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR