N-(2-Iodo-4,5-dimethoxyphenyl)acetamide ≥95%

Supplier: AOBChem USA

Warning

508500-5G 508500-25G 508500-1G 508500-500MG
77630-126EA 1602.68 USD
77630-126 77636-048 77643-504 77651-440
N-(2-Iodo-4,5-dimethoxyphenyl)acetamide ≥95%
N-(2-Iodo-4,5-dimethoxyphenyl)acetamide

Precaución: For Research Use Only

Formula: 321.1116 g/mol
MW: C₁₀H₁₂INO₃
Storage Temperature: Ambient
CAS Number: 550372-53-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR