(2,6-Dichloro-4-iodophenyl)(methyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

506393-5G 506393-1G 506393-500MG 506393-25G
77633-468EA 1441 USD
77633-468 77639-460 77652-976 77659-868
(2,6-Dichloro-4-iodophenyl)(methyl)sulfane ≥95%
(2,6-Dichloro-4-iodophenyl)(methyl)sulphane

Precaución: For Research Use Only

Formula: 318.9900 g/mol
MW: C₇H₅Cl₂IS
Storage Temperature: Ambient
CAS Number: 2711230-22-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR