4-Cyclopropyl-2-fluoro-3-methylpyridine ≥97%

Supplier: AOBChem USA

Warning

50573-5G 50573-25G 50573-1G 50573-500MG
77630-124EA 1652.12 USD
77630-124 77636-046 77639-456 77647-444
4-Cyclopropyl-2-fluoro-3-methylpyridine ≥97%
4-Cyclopropyl-2-fluoro-3-methylpyridine

Precaución: For Research Use Only

Formula: C₉H₁₀FN
Storage Temperature: Ambient
MDL Number: MFCD32902882
CAS Number: 2586126-09-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR