2-Bromo-3-fluoro-5-methylisonicotinic acid ≥97%

Supplier: AOBChem USA

Warning

504036-5G 504036-1G 504036-500MG
77630-122EA 1484.23 USD
77630-122 77639-454 77651-436
2-Bromo-3-fluoro-5-methylisonicotinic acid ≥97%
2-Bromo-3-fluoro-5-methylisonicotinic acid

Precaución: For Research Use Only

Formula: 234.0225 g/mol
MW: C₇H₅BrFNO₂
Storage Temperature: Ambient
CAS Number: 2932438-96-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR