1-(4-Bromophenyl)cyclobutane-1-carbonitrile ≥97%
Supplier: AOBChem USA
|
Warning
|
Synonyms:
1-(4-Bromophenyl)cyclobutanecarbonitrile
503856-5G
503856-25G
77635-122EA
68
USD
77635-122
77656-332
1-(4-Bromophenyl)cyclobutane-1-carbonitrile ≥97%
1-(4-Bromophenyl)cyclobutane-1-carbonitrile
Precaución: For Research Use Only
Formula:
C₁₁H₁₀BrN MW: 236.108 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD10688823 CAS Number: 485828-58-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...