3-Methoxy-5-(trifluoromethyl)benzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

50118-5G 50118-25G 50118-1G
77635-120EA 194 USD
77635-120 77656-328 77643-494
3-Methoxy-5-(trifluoromethyl)benzyl alcohol ≥97%
3-Methoxy-5-(trifluoromethyl)benzyl alcohol

Precaución: For Research Use Only

Formula: C₉H₉F₃O₂
Storage Temperature: Ambient
MDL Number: MFCD09025413
CAS Number: 916420-98-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR