4-Bromo-2-iodothiophene ≥97%

Supplier: AOBChem USA

Warning

50047-5G 50047-1G 50047-500MG
77659-866EA 1971 USD
77659-866 77643-492 77651-432
4-Bromo-2-iodothiophene ≥97%
4-Bromo-2-iodothiophene

Precaución: For Research Use Only

Formula: C₄H₂BrIS
Storage Temperature: Ambient
MDL Number: MFCD18451610
CAS Number: 73882-40-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR