5-Bromo-3-(trifluoromethyl)benzo[b]thiophene-2-carboxylic acid ≥97%

Supplier: AOBChem USA

Warning

500391-5G 500391-25G 500391-1G 500391-500MG
77630-120EA 1495.56 USD
77630-120 77636-044 77643-490 77651-430
5-Bromo-3-(trifluoromethyl)benzo[b]thiophene-2-carboxylic acid ≥97%
5-Bromo-3-(trifluoromethyl)benzo[b]thiophene-2-carboxylic acid

Precaución: For Research Use Only

Formula: C₁₀H₄BrF₃O₂S
MW: 325.102 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23115255
CAS Number: 826995-52-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR