Ethyl 5-bromo-3-(trifluoromethyl)benzo[b]thiophene-2-carboxylate ≥97%

Supplier: AOBChem USA

Warning

500390-5G 500390-25G 500390-1G 500390-500MG
77630-118EA 1484.23 USD
77630-118 77636-042 77643-488 77647-440
Ethyl 5-bromo-3-(trifluoromethyl)benzo[b]thiophene-2-carboxylate ≥97%
Ethyl 5-bromo-3-(trifluoromethyl)benzo[b]thiophene-2-carboxylate

Precaución: For Research Use Only

Formula: C₁₂H₈BrF₃O₂S
Storage Temperature: Ambient
CAS Number: 1522380-83-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR