Methyl 2,3-difluoro-6-methoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

50027-5G 50027-25G
77635-118EA 113 USD
77635-118 77656-326
Methyl 2,3-difluoro-6-methoxybenzoate ≥97%

Precaución: For Research Use Only

Formula: C₉H₈F₂O₃
Storage Temperature: Ambient
MDL Number: MFCD06204146
CAS Number: 773876-05-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR