2-Chloro-3-cyano-6-fluorobenzoic acid ≥95%

Supplier: AOBChem USA

Warning

49991-1G 49991-5G 49991-500MG
77639-450EA 751 USD
77639-450 77659-864 77651-428
2-Chloro-3-cyano-6-fluorobenzoic acid ≥95%
2-Chloro-3-cyano-6-fluorobenzoic acid

Precaución: For Research Use Only

Formula: 199.566 g/mol
MW: C₈H₃ClFNO₂
Storage Temperature: Ambient
CAS Number: 1805570-67-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR