N-(2-Hydroxyethyl)-3-methylbenzamide ≥97%

Supplier: AOBChem USA

Warning

499255-5G 499255-1G 499255-25G 499255-500MG
77633-464EA 1070 USD
77633-464 77639-446 77656-324 77647-438
N-(2-Hydroxyethyl)-3-methylbenzamide ≥97%
N-(2-Hydroxyethyl)-3-methylbenzamide

Precaución: For Research Use Only

Formula: C₁₀H₁₃NO₂
Storage Temperature: Ambient
CAS Number: 778623-19-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR