1-(3-Bromo-6-chloro-2-fluorophenyl)-2,2,2-trifluoroethanone ≥95%

Supplier: AOBChem USA

Warning

49910-1G 49910-5G 49910-500MG
77639-444EA 556 USD
77639-444 77656-322 77651-426
1-(3-Bromo-6-chloro-2-fluorophenyl)-2,2,2-trifluoroethanone ≥95%
1-(3-Bromo-6-chloro-2-fluorophenyl)-2,2,2-trifluoroethanone

Precaución: For Research Use Only

Formula: 305.451 g/mol
MW: C₈H₂BrClF₄O
Storage Temperature: Ambient
MDL Number: MFCD29914186
CAS Number: 943830-34-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR