(2,4-Dichloro-6-fluorophenyl)trimethylsilane ≥97%

Supplier: AOBChem USA

Warning

498671-5G 498671-500MG 498671-25G 498671-1G
77633-462EA 1441 USD
77633-462 77652-972 77659-860 77643-482
(2,4-Dichloro-6-fluorophenyl)trimethylsilane ≥97%
(2,4-Dichloro-6-fluorophenyl)trimethylsilane

Precaución: For Research Use Only

Formula: 237.174 g/mol
MW: C₉H₁₁Cl₂FSi
Storage Temperature: Ambient
CAS Number: 3037682-71-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR