Cyclopropyl(2,4-dichloro-6-fluorophenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

498670-5G 498670-25G 498670-1G 498670-500MG
77630-112EA 1484.23 USD
77630-112 77636-038 77643-480 77651-420
Cyclopropyl(2,4-dichloro-6-fluorophenyl)methanol ≥95%
Cyclopropyl(2,4-dichloro-6-fluorophenyl)methanol

Precaución: For Research Use Only

Formula: 235.082 g/mol
MW: C₁₀H₉Cl₂FO
Storage Temperature: Ambient
CAS Number: 3037705-36-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR