1-(2,4-Dichloro-6-fluorophenyl)-2,2,2-trifluoroethanone ≥97%

Supplier: AOBChem USA

Warning

498669-5G 498669-1G 498669-500MG 498669-25G
77630-110EA 1532.64 USD
77630-110 77639-442 77652-970 77656-318
1-(2,4-Dichloro-6-fluorophenyl)-2,2,2-trifluoroethanone ≥97%
1-(2,4-Dichloro-6-fluorophenyl)-2,2,2-trifluoroethanone

Precaución: For Research Use Only

Formula: 261 g/mol
MW: C₈H₂Cl₂F₄O
Storage Temperature: Ambient
CAS Number: 3037676-32-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR