1-(2,4-Dichloro-6-fluorophenyl)ethanol ≥95%

Supplier: AOBChem USA

Warning

498666-5G 498666-1G 498666-25G 498666-500MG
77630-108EA 1484.23 USD
77630-108 77639-440 77656-316 77651-418
1-(2,4-Dichloro-6-fluorophenyl)ethanol ≥95%
1-(2,4-Dichloro-6-fluorophenyl)ethanol

Precaución: For Research Use Only

Formula: 209.045 g/mol
MW: C₈H₇Cl₂FO
Storage Temperature: Ambient
CAS Number: 936645-58-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR