1-(2-Fluoro-3,4-dimethoxyphenyl)cyclobutanol ≥95%

Supplier: AOBChem USA

Warning

498665-5G 498665-1G 498665-25G 498665-500MG
77630-106EA 1484.23 USD
77630-106 77639-438 77659-858 77647-436
1-(2-Fluoro-3,4-dimethoxyphenyl)cyclobutanol ≥95%
1-(2-Fluoro-3,4-dimethoxyphenyl)cyclobutanol

Precaución: For Research Use Only

Formula: C₁₂H₁₅FO₃
Storage Temperature: Ambient
CAS Number: 3031812-73-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR