2,2,2-Trifluoro-1-(2-fluoro-3,4-dimethoxyphenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

498664-5G 498664-25G 498664-1G 498664-500MG
77630-104EA 1484.23 USD
77630-104 77656-314 77647-432 77647-434
2,2,2-Trifluoro-1-(2-fluoro-3,4-dimethoxyphenyl)ethanone ≥95%
2,2,2-Trifluoro-1-(2-fluoro-3,4-dimethoxyphenyl)ethanone

Precaución: For Research Use Only

Formula: 252.162 g/mol
MW: C₁₀H₈F₄O₃
Storage Temperature: Ambient
CAS Number: 3031789-04-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR