1-(6-(Tert-butylthio)-2-fluoropyridin-3-yl)ethanol ≥95%

Supplier: AOBChem USA

Warning

498646-5G 498646-1G 498646-25G 498646-500MG
77630-096EA 1484.23 USD
77630-096 77639-434 77656-310 77651-416
1-(6-(Tert-butylthio)-2-fluoropyridin-3-yl)ethanol ≥95%
1-(6-(Tert-butylthio)-2-fluoropyridin-3-yl)ethanol

Precaución: For Research Use Only

Formula: 229.314 g/mol
MW: C₁₁H₁₆FNOS
Storage Temperature: Ambient
CAS Number: 3031812-69-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR