(2-Methyl-4-(methylthio)phenyl)(phenyl)methanone ≥95%

Supplier: AOBChem USA

Warning

498636-5G 498636-1G 498636-25G 498636-500MG
77633-460EA 1441 USD
77633-460 77639-430 77659-854 77651-412
(2-Methyl-4-(methylthio)phenyl)(phenyl)methanone ≥95%
(2-Methyl-4-(methylthio)phenyl)(phenyl)methanone

Precaución: For Research Use Only

Formula: C₁₅H₁₄OS
Storage Temperature: Ambient
CAS Number: 3037682-72-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR