Cyclopropyl(2-methyl-4-(methylthio)phenyl)methanone ≥95%

Supplier: AOBChem USA

Warning

498635-5G 498635-25G 498635-1G 498635-500MG
77630-094EA 1484.23 USD
77630-094 77636-034 77643-474 77647-426
Cyclopropyl(2-methyl-4-(methylthio)phenyl)methanone ≥95%
Cyclopropyl(2-methyl-4-(methylthio)phenyl)methanone

Precaución: For Research Use Only

Formula: C₁₂H₁₄OS
Storage Temperature: Ambient
CAS Number: 3037695-54-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR