2,2,2-Trifluoro-1-(2-methyl-4-(methylthio)phenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

498634-5G 498634-25G 498634-1G 498634-500MG
77630-092EA 1484.23 USD
77630-092 77659-852 77643-472 77651-410
2,2,2-Trifluoro-1-(2-methyl-4-(methylthio)phenyl)ethanone ≥95%
2,2,2-Trifluoro-1-(2-methyl-4-(methylthio)phenyl)ethanone

Precaución: For Research Use Only

Formula: 234.238 g/mol
MW: C₁₀H₉F₃OS
Storage Temperature: Ambient
CAS Number: 3037703-45-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR