2-(2-Methyl-4-(methylthio)phenyl)propan-2-ol ≥95%

Supplier: AOBChem USA

Warning

498633-1G 498633-5G 498633-500MG
77639-428EA 583 USD
77639-428 77659-850 77647-424
2-(2-Methyl-4-(methylthio)phenyl)propan-2-ol ≥95%
2-(2-Methyl-4-(methylthio)phenyl)propan-2-ol

Precaución: For Research Use Only

Formula: 196.309 g/mol
MW: C₁₁H₁₆OS
Storage Temperature: Ambient
CAS Number: 1896661-73-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR