Tert-butyl 2-methoxy-3-methylbenzoate ≥95%

Supplier: AOBChem USA

Warning

498625-5G 498625-25G 498625-1G 498625-500MG
77630-088EA 1484.23 USD
77630-088 77636-032 77643-464 77651-402
Tert-butyl 2-methoxy-3-methylbenzoate ≥95%
Tert-butyl 2-methoxy-3-methylbenzoate

Precaución: For Research Use Only

Formula: C₁₃H₁₈O₃
Storage Temperature: Ambient
CAS Number: 2377377-83-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR