2-(2-Methoxy-3-methylphenyl)propan-2-ol ≥95%

Supplier: AOBChem USA

Warning

498622-5G 498622-500MG 498622-25G 498622-1G
77633-456EA 1171 USD
77633-456 77652-964 77656-306 77643-460
2-(2-Methoxy-3-methylphenyl)propan-2-ol ≥95%
2-(2-Methoxy-3-methylphenyl)propan-2-ol

Precaución: For Research Use Only

Formula: 180.244 g/mol
MW: C₁₁H₁₆O₂
Storage Temperature: Ambient
CAS Number: 850565-13-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR